Assay Details
Target Gene Details
Entrez Gene ID: | 75104 |
Gene Name: | monocyte to macrophage differentiation-associated 2 |
Gene Aliases: |
4930518M15Rik, C88001 |
Location: |
Chr.5:142563474-142608814 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mmd2 | NM_175217.6 | 7 | 931 | NP_780426.1 |
| AY134664.1 | 7 | 872 | AAN08615.1 | |
| AY424299.1 | 7 | 931 | AAR08387.1 | |
| BC025064.1 | 7 | 931 | AAH25064.1 |
More Information
Set Membership: |
|

Genomic Map