Assay Details
Target Gene Details
Entrez Gene ID: | 78937 |
Gene Name: | AVL9 homolog (S. cerevisiase) |
Gene Aliases: |
5830411G16Rik, D730049P16Rik, mKIAA0241 |
Location: |
Chr.6:56714905-56761912 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Avl9 | NM_030235.1 | 1 | 105 | NP_084511.1 |
| AK020003.1 | 1 | 82 | ||
| AK083605.1 | 1 | 61 | ||
| AK122228.1 | 1 | 83 | BAC65510.1 | |
| AK147365.1 | 1 | 107 | ||
| BC117878.1 | 1 | 67 | ||
| CJ109208.1 | 1 | 107 |

Genomic Map