Assay Details
Target Gene Details
Entrez Gene ID: | 51811 |
Gene Name: | C-type lectin domain family 4, member f |
Gene Aliases: |
Clecsf13, D18063, KUCR_MOUSE, Kclr |
Location: |
Chr.6:83644542-83656116 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clec4f | NM_016751.3 | 7 | 2268 | NP_058031.2 |
| AK040696.1 | 7 | 2269 | BAC30671.1 |
More Information
Set Membership: |
|

Genomic Map