Assay Details
Target Gene Details
Entrez Gene ID: | 27056 |
Gene Name: | interferon regulatory factor 5 |
Gene Aliases: |
AW491843, mirf5 |
Location: |
Chr.6:29526130-29541851 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Irf5 | NM_001252382.1 | 7 | 1135 | NP_001239311.1 |
| NM_012057.4 | 7 | 1083 | NP_036187.1 | |
| AF028725.1 | 7 | 975 | AAB81997.1 | |
| AK150598.1 | 7 | 1002 | ||
| AK151019.1 | 7 | 1000 | BAE30037.1 | |
| AK152042.1 | 7 | 1002 | ||
| AK156220.1 | 7 | 1135 | BAE33631.1 | |
| AK171356.1 | 7 | 1003 | ||
| EU401974.1 | 4 | 634 |
More Information
Set Membership: |
|

Genomic Map