Assay Details
Target Gene Details
Entrez Gene ID: | 57905 |
Gene Name: | ISY1 splicing factor homolog (S. cerevisiae) |
Gene Aliases: |
5830446M03Rik, AI181014, AU020769 |
Location: |
Chr.6:87818447-87838759 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Isy1 | NM_133934.4 | 11 | 1117 | NP_598695.1 |
| AK017996.1 | 11 | 1034 | BAB31028.1 | |
| BC020103.1 | 11 | 1081 | AAH20103.1 | |
| BC022597.1 | 4 | 589 | AAH22597.1 | |
| BC037695.1 | 11 | 1122 | AAH37695.1 |
More Information
Set Membership: |
|

Genomic Map