Assay Details
Target Gene Details
Entrez Gene ID: | 68929 |
Gene Name: | motile sperm domain containing 3 |
Gene Aliases: |
1190005J19Rik, 5133401H10Rik, Gtig2, R124 |
Location: |
Chr.5:137596645-137601083 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mospd3 | NM_030037.2 | 1 | 68 | NP_084313.1 |
| BC039620.1 | 1 | 49 | AAH39620.1 |

Genomic Map