Assay Details
Target Gene Details
Entrez Gene ID: | 57252 |
Gene Name: | taste receptor, type 2, member 105 |
Gene Aliases: |
T2R05, T2R9, T2r5, Tas2r5, mGR05, mt2r5 |
Location: |
Chr.6:131686561-131687463 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tas2r105 | NM_020501.1 | 1 | 305 | NP_065247.1 |
| BC125515.1 | 1 | 337 | ||
| BC125541.1 | 1 | 337 |
More Information
Set Membership: |
|

Genomic Map