Assay Details
Target Gene Details
Entrez Gene ID: | 387345 |
Gene Name: | taste receptor, type 2, member 113 |
Gene Aliases: |
T2R13, Tas2r13, mGR13, mt2r58 |
Location: |
Chr.6:132893011-132893940 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tas2r113 | NM_207018.1 | 1 | 155 | NP_996901.1 |
More Information
Set Membership: |
|

Genomic Map