Assay Details
Target Gene Details
Entrez Gene ID: | 387346 |
Gene Name: | taste receptor, type 2, member 114 |
Gene Aliases: |
T2R14, Tas2r14, mGR14, mt2r46 |
Location: |
Chr.6:131689134-131690063 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tas2r114 | NM_207019.1 | 1 | 460 | NP_996902.1 |
More Information
Set Membership: |
|

Genomic Map