Assay Details
Target Gene Details
Entrez Gene ID: | 26433 |
Gene Name: | procollagen-lysine, 2-oxoglutarate 5-dioxygenase 3 |
Gene Aliases: |
AI414586, LH3 |
Location: |
Chr.5:136987019-136996646 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plod3 | NM_011962.3 | 1 | 137 | NP_036092.1 |
| BC043047.1 | 1 | 134 | AAH43047.1 | |
| BC054734.1 | 1 | 137 | AAH54734.1 |
Target Gene Details
Entrez Gene ID: | 70103 |
Gene Name: | zinc finger, HIT domain containing 1 |
Gene Aliases: |
2700001K05Rik, ZNFN4A1 |
Location: |
Chr.5:136982194-136987882 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Znhit1 | NM_027318.3 | NP_081594.1 | ||
| AK012199.1 | BAB28094.1 | |||
| AK080799.1 | BAC38025.1 |
More Information
Set Membership: |
|

Genomic Map