Assay Details
Target Gene Details
Entrez Gene ID: | 20349 |
Gene Name: | sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3E |
Gene Aliases: |
6430702L12, AA408817, Semah, mKIAA0331 |
Location: |
Chr.5:14025276-14256689 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sema3e | NM_011348.2 | 1 | 496 | NP_035478.2 |
| AF034744.1 | 1 | 231 | AAD01996.1 | |
| AK141295.1 | 1 | 497 | ||
| AK142340.1 | 1 | 323 | BAE25038.1 | |
| AK159214.1 | 1 | 287 | BAE34903.1 | |
| BC057956.1 | 1 | 394 | AAH57956.1 | |
| Z93947.1 | 1 | 468 | CAB07987.1 | |
| Z93948.1 | 1 | 468 | CAB07988.1 |
More Information
Set Membership: |
|

Genomic Map