Assay Details
Target Gene Details
Entrez Gene ID: | 78412 |
Gene Name: | RIKEN cDNA 3110062M04 gene |
Gene Aliases: |
AL023049, Mri |
Location: |
Chr.6:34871771-34879019 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 3110062M04Rik | NM_001135611.1 | NP_001129083.1 | ||
| NM_199145.2 | 1 | 254 | NP_954596.1 | |
| AK019439.1 | 1 | 190 | ||
| AK042380.1 | 1 | 210 | BAC31243.1 | |
| AK083365.1 | ||||
| AK170119.1 | BAE41576.1 | |||
| BB611813.1 | ||||
| BC029235.1 | AAH29235.1 |

Genomic Map