Assay Details
Target Gene Details
Entrez Gene ID: | 79362 |
Gene Name: | basic helix-loop-helix family, member e41 |
Gene Aliases: |
6430520M22Rik, Bhlhb2l, Bhlhb3, DEC2, Sharp1 |
Location: |
Chr.6:145858243-145865591 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bhlhe41 | NM_001271768.1 | 5 | 956 | NP_001258697.1 |
| NM_024469.2 | 5 | 827 | NP_077789.1 | |
| AB044090.1 | 5 | 747 | BAB21503.1 | |
| AK137275.1 | 1 | 116 |
More Information
Set Membership: |
|

Genomic Map