Assay Details
Target Gene Details
Entrez Gene ID: | 320609 |
Gene Name: | striatin interacting protein 2 |
Gene Aliases: |
D330017J20Rik, Fam40b |
Location: |
Chr.6:29916976-29959681 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Strip2 | NM_001037740.1 | 21 | 2772 | NP_001032829.1 |
| NM_177204.3 | 21 | 2856 | NP_796178.2 | |
| NR_104576.1 | 21 | 2852 | ||
| AK034933.1 | ||||
| AK040556.1 | 1 | 135 | ||
| AK042073.1 | 21 | 2857 | BAC31154.1 | |
| AK052271.1 | 20 | 3180 | ||
| AK052273.1 | 13 | 1882 | ||
| AK122459.1 | 21 | 2577 | BAC65741.1 | |
| BC147502.1 |

Genomic Map