Assay Details
Target Gene Details
Entrez Gene ID: | 668210 |
Gene Name: | predicted gene 9047 |
Gene Aliases: |
EG668210 |
Location: |
Chr.6:29471437-29473429 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm9047 | NM_001145360.1 | 1 | 251 | NP_001138832.1 |
| BC147635.1 | 1 | 251 | ||
| BC147645.1 | 1 | 251 |

Genomic Map