Assay Details
Target Gene Details
Entrez Gene ID: | 16485 |
Gene Name: | potassium voltage-gated channel, shaker-related subfamily, member 1 |
Gene Aliases: |
AI840627, Kca1-1, Kv1.1, MBK1, Mk-1, Shak, mceph |
Location: |
Chr.6:126636463-126645801 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcna1 | NM_010595.3 | 1 | 265 | NP_034725.3 |
More Information
Set Membership: |
|

Genomic Map