Assay Details
Target Gene Details
Entrez Gene ID: | 19362 |
Gene Name: | RAD51 associated protein 1 |
Gene Aliases: |
2510006L10Rik, RAB22 |
Location: |
Chr.6:126923415-126939621 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rad51ap1 | NM_009013.3 | 9 | 1226 | NP_033039.2 |
| AK010920.1 | 7 | 1225 | ||
| AK011379.1 | 9 | 1227 | BAB27579.1 | |
| AK077829.1 | 9 | 1256 | BAC37025.1 | |
| AK079525.1 | 9 | 1248 | BAC37671.1 | |
| BC003738.1 | 9 | 1240 | AAH03738.1 |

Genomic Map