Assay Details
Target Gene Details
Entrez Gene ID: | 112408 |
Gene Name: | taste receptor, type 2, member 116 |
Gene Aliases: |
T2R16, TRB1, TRB4, Tas2r14, Tas2r16, Tas2r7, mGR16, mt2r56 |
Location: |
Chr.6:132855438-132856355 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tas2r116 | NM_053212.1 | 1 | 762 | NP_444442.1 |
| AF247734.1 | 1 | 669 | AAF64511.1 |
More Information
Set Membership: |
|

Genomic Map