Assay Details
Target Gene Details
Entrez Gene ID: | 387348 |
Gene Name: | taste receptor, type 2, member 120 |
Gene Aliases: |
T2R20, Tas2r20, mGR20, mt2r47 |
Location: |
Chr.6:132656957-132657844 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tas2r120 | NM_207023.1 | 1 | 445 | NP_996906.1 |
More Information
Set Membership: |
|

Genomic Map