Assay Details
Target Gene Details
Entrez Gene ID: | 387349 |
Gene Name: | taste receptor, type 2, member 121 |
Gene Aliases: |
T2R21, Tas2r13, Tas2r21, mGR21, mT2r48 |
Location: |
Chr.6:132700090-132701007 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tas2r121 | NM_207024.1 | 1 | 765 | NP_996907.1 |
| BC104410.1 | 1 | 765 | AAI04411.1 | |
| BC104411.1 | 1 | 765 | AAI04412.1 |
More Information
Set Membership: |
|

Genomic Map