Assay Details
Target Gene Details
Entrez Gene ID: | 53620 |
Gene Name: | vesicle-associated membrane protein 5 |
Gene Aliases: |
AF119384, Camp |
Location: |
Chr.6:72368049-72380468 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vamp5 | NM_001080742.2 | 4 | 938 | NP_001074211.1 |
| NM_016872.4 | 3 | 1048 | NP_058568.1 | |
| AF119384.1 | AAD29365.1 | |||
| BC145146.1 |

Genomic Map