Assay Details
Target Gene Details
Entrez Gene ID: | 67054 |
Gene Name: | phosphoribosylaminoimidazole carboxylase, phosphoribosylaminoribosylaminoimidazole, succinocarboxamide synthetase |
Gene Aliases: |
2610511I09Rik, ADE2H1, AIRC, PAIS |
Location: |
Chr.5:76951315-76967507 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Paics | NM_025939.2 | 1 | 62 | NP_080215.1 |
| AK146722.1 | 1 | 114 | BAE27385.1 | |
| BC052691.1 | 1 | 62 | AAH52691.1 |
Target Gene Details
Entrez Gene ID: | 231327 |
Gene Name: | phosphoribosyl pyrophosphate amidotransferase |
Gene Aliases: |
5730454C12Rik, AA408689, AA675351, AI507113, C79945 |
Location: |
Chr.5:76913249-76951578 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppat | NM_172146.2 | 1 | 107 | NP_742158.1 |
| AK159141.1 | 1 | 101 | ||
| AK159859.1 | 1 | 101 | ||
| BB647933.1 | 1 | 107 |
More Information
Set Membership: |
|

Genomic Map