Assay Details
Target Gene Details
Entrez Gene ID: | 108014 |
Gene Name: | serine/arginine-rich splicing factor 9 |
Gene Aliases: |
25kDa, 2610029M16Rik, SRp30c, Sfrs9 |
Location: |
Chr.5:115327161-115333084 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srsf9 | NM_025573.3 | 4 | 1076 | NP_079849.1 |
| NR_036616.1 | 5 | 1166 | ||
| AI503042.1 | 1 | 94 | ||
| AK011621.1 | 4 | 1036 | BAB27740.1 | |
| AK020084.1 | 4 | 848 | BAB31986.1 | |
| AK159190.1 | 4 | 1074 | BAE34886.1 | |
| BC012217.1 | 4 | 1076 | AAH12217.1 | |
| CV558546.1 |
More Information
Set Membership: |
|

Genomic Map