Assay Details
Target Gene Details
Entrez Gene ID: | 64654 |
Gene Name: | fibroblast growth factor 23 |
Gene Aliases: |
- |
Location: |
Chr.6:127072902-127082296 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fgf23 | NM_022657.4 | 1 | 55 | NP_073148.1 |
| AF263536.1 | AAG09916.1 | |||
| AK155687.1 | 1 | 55 | BAE33388.1 | |
| BC120605.1 |
More Information
Set Membership: |
|

Genomic Map