Assay Details
Target Gene Details
Entrez Gene ID: | 387616 |
Gene Name: | taste receptor, type 2, member 140 |
Gene Aliases: |
T2R40, T2r34, TRB3, TRB5, Tas2r13, Tas2r137, Tas2r40, Tas2r8, mTRB3, mt2r64 |
Location: |
Chr.6:133054855-133055793 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tas2r140 | NM_021562.1 | 1 | 629 | NP_067537.1 |
| AF247735.1 | 1 | 536 | AAF64512.1 | |
| BC145965.1 | 1 | 652 | ||
| BC145967.1 | 1 | 652 |
More Information
Set Membership: |
|

Genomic Map