Assay Details
Target Gene Details
Entrez Gene ID: | 79264 |
Gene Name: | KRIT1, ankyrin repeat containing |
Gene Aliases: |
2010007K12Rik, A630036P20Rik, AA432855, AI450393, AI643869, BB155247, BB235701, Ccm1 |
Location: |
Chr.5:3803165-3844515 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krit1 | NM_001170552.1 | 1 | 177 | NP_001164023.1 |
| NM_030675.3 | 1 | 177 | NP_109600.2 | |
| NR_033173.1 | 1 | 161 | ||
| AF310134.1 | 1 | 153 | AAG47775.1 | |
| AK008150.1 | 1 | 159 | ||
| AK082175.1 | 1 | 159 | ||
| AK085107.1 | 1 | 159 | ||
| AK088956.1 | 1 | 162 | ||
| AK144667.1 | 1 | 159 | ||
| AK172215.1 | 1 | 159 | ||
| BB591998.1 | 1 | 172 | ||
| BB619038.1 | 1 | 159 | ||
| BB853405.1 | 1 | 160 | ||
| BB855141.1 | 1 | 177 | ||
| BC054819.1 | 1 | 153 | AAH54819.1 | |
| BC141042.1 | ||||
| BY150935.1 | 1 | 159 |
More Information
Set Membership: |
|

Genomic Map