Assay Details
Target Gene Details
Entrez Gene ID: | 623131 |
Gene Name: | proline rich 19 |
Gene Aliases: |
EG623131 |
Location: |
Chr.7:25301359-25304140 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prr19 | NM_001081294.1 | 3 | 833 | NP_001074763.1 |
| BC147639.1 | 3 | 781 | ||
| BC147641.1 | 3 | 781 | ||
| BC147701.1 | 3 | 781 | ||
| BC147708.1 | 3 | 781 |

Genomic Map