Assay Details
Target Gene Details
Entrez Gene ID: | 67893 |
Gene Name: | transmembrane protein 86A |
Gene Aliases: |
1810054O13Rik, AI414959, AW742698 |
Location: |
Chr.7:47050640-47054776 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem86a | NM_026436.3 | 3 | 1335 | NP_080712.1 |
| AK007864.1 | 3 | 1336 | BAB25316.1 | |
| AK075589.1 | 3 | 1309 | BAC35840.1 | |
| AK213808.1 | 1 | 312 | ||
| BC056619.1 | 3 | 1302 | AAH56619.1 |

Genomic Map