Assay Details
Target Gene Details
Entrez Gene ID: | 244237 |
Gene Name: | tumor necrosis factor receptor superfamily, member 26 |
Gene Aliases: |
Tnfrh3, Tnfrsf24 |
Location: |
Chr.7:143607685-143627845 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfrsf26 | NM_175649.5 | 8 | 1003 | NP_783580.1 |
| AK154666.1 | 8 | 1114 | BAE32753.1 | |
| AY165628.1 | 8 | 1003 | AAN87808.1 | |
| BC138819.1 | 8 | 1052 | ||
| BC138820.1 | 8 | 1053 |
More Information
Set Membership: |
|

Genomic Map