Assay Details
Target Gene Details
Entrez Gene ID: | 68332 |
Gene Name: | succinate dehydrogenase complex assembly factor 1 |
Gene Aliases: |
0610010E21Rik, AI430885, AW490662, Lyrm8 |
Location: |
Chr.7:30321409-30322375 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sdhaf1 | NM_001033140.3 | 1 | 648 | NP_001028312.2 |
| AA266979.1 | 1 | 176 | ||
| AK002462.1 | 1 | 632 | ||
| AK155437.1 | 1 | 612 | BAE33266.1 | |
| CA451543.1 | 1 | 620 |
More Information
Set Membership: |
|

Genomic Map