Assay Details
Target Gene Details
Entrez Gene ID: | 20646 |
Gene Name: | small nuclear ribonucleoprotein N |
Gene Aliases: |
2410045I01Rik, HCERN3, Peg4, Pwcr1, SMN |
Location: |
Chr.7:59982502-60140219 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snrpn | NM_001082961.1 | 11 | 1382 | NP_001076430.1 |
| NM_001082962.1 | 11 | 1426 | NP_001076431.1 | |
| NM_013670.3 | 10 | 1320 | NP_038698.1 | |
| AK010671.1 | ||||
| AK013607.1 | BAB28927.1 | |||
| AK144386.1 | 10 | 1276 | BAE25860.1 | |
| BC019589.1 | AAH19589.1 | |||
| BC024880.1 | AAH24880.1 | |||
| BU708754.1 | 4 | 523 | ||
| CD775913.1 | ||||
| X62648.1 | CAA44517.1 | |||
| X63730.1 | CAA45273.1 |
Target Gene Details
Entrez Gene ID: | 84704 |
Gene Name: | SNRPN upstream reading frame |
Gene Aliases: |
2410045I01Rik, Snrpn |
Location: |
Chr.7:59982501-60005156 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snurf | NM_033174.3 | 10 | 1320 | NP_149409.1 |
| BY648312.1 | ||||
| CD774269.1 | ||||
| CD776984.1 |
More Information
Set Membership: |
|

Genomic Map