Assay Details
Target Gene Details
Entrez Gene ID: | 233071 |
Gene Name: | Rho GTPase activating protein 33 |
Gene Aliases: |
AW324378, NOMA-GAP, Snx26, TCGAP |
Location: |
Chr.7:30522226-30535092 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap33 | NM_001289670.1 | 21 | 4068 | NP_001276599.1 |
| NM_001289682.1 | 22 | 4151 | NP_001276611.1 | |
| NM_178252.3 | 22 | 4178 | NP_839983.1 | |
| AK142180.1 | 21 | 4069 | BAE24965.1 | |
| BC061471.1 | 15 | 3355 | AAH61471.1 | |
| BC065086.1 | 22 | 4127 | AAH65086.1 | |
| BC065166.1 | 22 | 4125 | AAH65166.1 | |
| BC066047.1 | 22 | 4103 | AAH66047.1 |
More Information
Set Membership: |
|

Genomic Map