Assay Details
Target Gene Details
Entrez Gene ID: | 53333 |
Gene Name: | translocase of outer mitochondrial membrane 40 homolog (yeast) |
Gene Aliases: |
AW539759, Mom35, Tom40 |
Location: |
Chr.7:19701313-19715429 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tomm40 | NM_001109748.1 | 1 | 154 | NP_001103218.1 |
| NM_016871.2 | NP_058567.2 | |||
| AF043249.1 | AAC82341.1 | |||
| AF316403.1 | AAL46628.1 | |||
| AF316404.1 | AAL46629.1 | |||
| AK150019.1 | 1 | 151 | BAE29244.1 | |
| AK163759.1 | BAE37483.1 | |||
| AK164222.1 | ||||
| AK164570.1 | BAE37837.1 | |||
| AK165173.1 | BAE38058.1 | |||
| AK170991.1 | BAE42165.1 | |||
| BC100452.1 | AAI00453.1 | |||
| BY231811.1 |
More Information
Set Membership: |
|

Genomic Map