Assay Details
Target Gene Details
Entrez Gene ID: | 233913 |
Gene Name: | cDNA sequence BC017158 |
Gene Aliases: |
- |
Location: |
Chr.7:128271379-128298165 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BC017158 | NM_145590.2 | 1 | 223 | NP_663565.2 |
| AK148236.1 | 1 | 214 | BAE28428.1 | |
| AK149645.1 | 1 | 202 | BAE29003.1 | |
| AK154566.1 | 1 | 189 | ||
| AK164488.1 | 1 | 203 | ||
| AK165287.1 | 1 | 202 | ||
| AK170611.1 | 1 | 202 | BAE41911.1 | |
| BC017158.1 | 1 | 205 | AAH17158.1 | |
| BY236419.1 | 1 | 263 |

Genomic Map