Assay Details
Target Gene Details
Entrez Gene ID: | 233913 |
Gene Name: | cDNA sequence BC017158 |
Gene Aliases: |
- |
Location: |
Chr.7:128271379-128298165 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BC017158 | NM_145590.2 | 13 | 2181 | NP_663565.2 |
| AK148236.1 | 13 | 2172 | BAE28428.1 | |
| AK149645.1 | 13 | 2160 | BAE29003.1 | |
| AK154566.1 | 13 | 2149 | ||
| AK164488.1 | 13 | 2159 | ||
| AK210322.1 | ||||
| BC017158.1 | 13 | 2163 | AAH17158.1 | |
| BP764390.1 | 1 | 445 |
Target Gene Details
Entrez Gene ID: | 246787 |
Gene Name: | solute carrier family 5 (sodium/glucose cotransporter), member 2 |
Gene Aliases: |
Sglt2 |
Location: |
Chr.7:128265697-128272433 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc5a2 | NM_133254.3 | NP_573517.1 | ||
| AK128900.1 | ||||
| AY033886.1 | AAK61606.1 | |||
| BC022226.1 | AAH22226.1 |
More Information
Set Membership: |
|

Genomic Map