Assay Details
Target Gene Details
Entrez Gene ID: | 170639 |
Gene Name: | olfactory receptor 78 |
Gene Aliases: |
4633402A21Rik, MOL2.3, MOR18-2, Or51e2, PSGR, RA1c |
Location: |
Chr.7:102740721-102759471 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olfr78 | NM_001168503.1 | 3 | 684 | NP_001161975.1 |
| NM_130866.4 | 4 | 1553 | NP_570936.1 | |
| AF378854.1 | 1 | 338 | AAL35109.1 | |
| AK028467.1 | 4 | 1478 | BAC25966.1 | |
| AK036356.1 | 3 | 685 | BAC29396.1 | |
| BC120603.1 | 2 | 527 | ||
| BC120629.1 | 2 | 527 |
More Information
Set Membership: |
|

Genomic Map