Assay Details
Target Gene Details
Entrez Gene ID: | 100042056 |
Gene Name: | RIKEN cDNA 9130019P16 gene |
Gene Aliases: |
6430501H15Rik, OTTMUSG00000023151 |
Location: |
Chr.6:54269681-54430221 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 9130019P16Rik | NR_033635.1 | 1 | 548 | |
| AK032212.1 | 1 | 548 | BAC27762.1 |
Target Gene Details
Entrez Gene ID: | 330319 |
Gene Name: | WAS/WASL interacting protein family, member 3 |
Gene Aliases: |
9030621G03, BQ952480 |
Location: |
Chr.6:54429218-54503768 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wipf3 |

Genomic Map