Assay Details
Target Gene Details
Entrez Gene ID: | 73813 |
Gene Name: | family with sequence similarity 83, member E |
Gene Aliases: |
4930403C10Rik |
Location: |
Chr.7:45721220-45729492 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam83e | NM_001033170.4 | NP_001028342.2 | ||
| BC042793.1 | AAH42793.1 |
Target Gene Details
Entrez Gene ID: | 69363 |
Gene Name: | sperm acrosome associated 4 |
Gene Aliases: |
1700008E09Rik, AV043694, Samp14 |
Location: |
Chr.7:45725107-45725816 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spaca4 | NM_027055.3 | 1 | 120 | NP_081331.2 |
| AK005760.1 | 1 | 112 | BAB24224.1 | |
| BC048608.1 | 1 | 117 | AAH48608.1 |
Target Gene Details
Entrez Gene ID: | 54200 |
Gene Name: | sulfotransferase family, cytosolic, 2B, member 1 |
Gene Aliases: |
AI326997, BB173635, ST2B1, SULT2B |
Location: |
Chr.7:45721174-45759555 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sult2b1 |
More Information
Set Membership: |
|

Genomic Map