Assay Details
Target Gene Details
Entrez Gene ID: | 56632 |
Gene Name: | sphingosine kinase 2 |
Gene Aliases: |
C76851 |
Location: |
Chr.7:45709463-45718002 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sphk2 | NM_001172561.1 | 2 | 458 | NP_001166032.1 |
| NM_020011.5 | 2 | 767 | NP_064395.2 | |
| NM_203280.3 | 3 | 529 | NP_975009.1 | |
| AF245448.1 | 3 | 479 | AAF74125.1 | |
| AF415214.1 | 1 | 93 | AAL07500.1 | |
| AK004951.1 | 2 | 767 | BAB23694.1 | |
| AK146414.1 | 2 | 360 | ||
| AK151990.1 | 3 | 429 | BAE30855.1 | |
| AK154321.1 | 2 | 195 | BAE32512.1 | |
| AK200767.1 | ||||
| AK206566.1 | ||||
| BC006941.1 | 2 | 201 | AAH06941.1 | |
| BC053737.1 | 3 | 442 | AAH53737.1 | |
| BC092084.1 | 2 | 216 | AAH92084.1 |
More Information
Set Membership: |
|

Genomic Map