Assay Details
Target Gene Details
Entrez Gene ID: | 12751 |
Gene Name: | tripeptidyl peptidase I |
Gene Aliases: |
Cln2, TPP-I |
Location: |
Chr.7:105744847-105752282 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tpp1 | NM_009906.5 | 13 | 3172 | NP_034036.1 |
| AK048279.1 | 13 | 3173 | BAC33293.1 | |
| AK155021.1 | 13 | 3191 | BAE32994.1 | |
| AK165669.1 | 13 | 3198 | ||
| AK166648.1 | 11 | 2965 | ||
| AK170781.1 | 13 | 3168 | BAE42026.1 |
More Information
Set Membership: |
|

Genomic Map