Assay Details
Target Gene Details
Entrez Gene ID: | 73919 |
Gene Name: | LYR motif containing 1 |
Gene Aliases: |
1110065L10Rik, 2310004B22Rik, 4930404J24Rik |
Location: |
Chr.7:119895807-119956203 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lyrm1 | AK015083.1 | BAB29706.1 | ||
| BY752674.1 |

Genomic Map