Assay Details
Target Gene Details
Entrez Gene ID: | 69387 |
Gene Name: | DnaJ (Hsp40) related, subfamily B, member 13 |
Gene Aliases: |
1700014P03Rik, Tsarg |
Location: |
Chr.7:100503011-100514815 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajb13 | NM_153527.2 | 8 | 955 | NP_705755.2 |
| AF419292.2 | 8 | 1100 | AAN32703.2 | |
| AK005984.1 | 8 | 972 | ||
| BC048501.1 | 8 | 955 | AAH48501.1 |
More Information
Set Membership: |
|

Genomic Map