Assay Details
Target Gene Details
Entrez Gene ID: | 80749 |
Gene Name: | leucine rich repeat and fibronectin type III domain containing 1 |
Gene Aliases: |
BC004018, SALM2, Semo1 |
Location: |
Chr.7:28451985-28468242 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrfn1 | NM_001141921.1 | 5 | 1779 | NP_001135393.1 |
| AB243740.1 | 2 | 1561 | BAE53711.1 | |
| DQ070870.1 | 2 | 1561 | AAZ20639.1 | |
| DQ314497.1 | 2 | 1561 |

Genomic Map