Assay Details
Target Gene Details
Entrez Gene ID: | 69731 |
Gene Name: | gem (nuclear organelle) associated protein 7 |
Gene Aliases: |
2400008I04Rik, AI120175, AI461688 |
Location: |
Chr.7:19564949-19573343 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gemin7 | NM_027189.2 | 2 | 162 | NP_081465.1 |
| AK010303.1 | 2 | 164 | BAB26836.1 | |
| BC028527.1 | 2 | 158 | AAH28527.1 |

Genomic Map