Assay Details
Target Gene Details
Entrez Gene ID: | 233067 |
Gene Name: | leucine rich repeat and fibronectin type III domain containing 3 |
Gene Aliases: |
A530045B06Rik, SALM4 |
Location: |
Chr.7:30355514-30362772 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrfn3 | NM_175478.2 | 3 | 2763 | NP_780687.1 |
| AK028115.1 | 2 | 571 | BAC25755.1 | |
| AK040916.1 | 3 | 2764 | BAC30743.1 | |
| BC066999.1 | 4 | 2686 | AAH66999.1 | |
| BC094593.1 | 3 | 2725 | AAH94593.1 | |
| DQ078787.1 | AAZ23616.1 |
More Information
Set Membership: |
|

Genomic Map