Assay Details
Target Gene Details
Entrez Gene ID: | 378430 |
Gene Name: | nanos homolog 2 (Drosophila) |
Gene Aliases: |
nos2 |
Location: |
Chr.7:18987524-18988962 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nanos2 | NM_194064.2 | 1 | 240 | NP_918953.2 |
| AB095972.1 | 1 | 240 | BAC82557.1 | |
| JQ936992.1 | 1 | 170 |
More Information
Set Membership: |
|

Genomic Map