Assay Details
Target Gene Details
Entrez Gene ID: | 76718 |
Gene Name: | catsper channel auxiliary subunit gamma 2 |
Gene Aliases: |
1700067C01Rik, CATSPERG |
Location: |
Chr.7:29697217-29727048 on Build GRCm38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Catsperg2 | NM_029714.3 | 29 | 3519 | NP_083990.2 |
| AK077059.1 | 26 | 4018 | ||
| AV277342.2 | 1 | 235 | ||
| BU938829.1 | 4 | 487 | ||
| GQ225581.1 |
More Information
Set Membership: |
|

Genomic Map