Assay Details
Target Gene Details
Entrez Gene ID: | 232946 |
Gene Name: | biogenesis of lysosomal organelles complex-1, subunit 3 |
Gene Aliases: |
BC043666, Blos3, E230011O18, rp |
Location: |
Chr.7:19505804-19508371 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bloc1s3 | NM_177692.3 | 2 | 630 | NP_808360.2 |
| AK054006.1 | 2 | 649 | BAC35618.1 | |
| BC025913.1 | 1 | 420 | AAH25913.1 | |
| BC043666.1 | 2 | 630 | AAH43666.1 |

Genomic Map