Assay Details
Target Gene Details
Entrez Gene ID: | 233016 |
Gene Name: | biliverdin reductase B (flavin reductase (NADPH)) |
Gene Aliases: |
- |
Location: |
Chr.7:27447978-27465981 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Blvrb | NM_001290525.1 | 5 | 607 | NP_001277454.1 |
| NM_144923.3 | 5 | 693 | NP_659172.1 | |
| AK153153.1 | 5 | 614 | BAE31763.1 | |
| AK164759.1 | BAE37903.1 | |||
| BC006617.1 | 5 | 633 | AAH06617.1 | |
| BC027279.1 | 5 | 693 | AAH27279.1 |
More Information
Set Membership: |
|

Genomic Map