Assay Details
Target Gene Details
Entrez Gene ID: | 57296 |
Gene Name: | proteasome (prosome, macropain) 26S subunit, non-ATPase, 8 |
Gene Aliases: |
6720456J22Rik, AA407360, AL033291, AL033322, AL033323, C76433 |
Location: |
Chr.7:29174187-29180673 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psmd8 | NM_026545.3 | 1 | 133 | NP_080821.3 |
| AK002921.1 | BAB22458.2 | |||
| AK003436.1 | BAB22789.2 | |||
| AK020132.1 | BAB32006.2 | |||
| AK051257.1 | BAC34576.1 | |||
| AK150493.1 | BAE29609.1 | |||
| AK153329.1 | BAE31908.1 | |||
| CF732286.1 | 1 | 133 |
More Information
Set Membership: |
|

Genomic Map